Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ced-12
Entrez Gene ID
34633
Gene Name
CG5336-PA
Gene Aliases
CG5336, Ced12, Dced-12, Dmel\CG5336, Dmel_CG5336, ELMO, Elmo, ced-12, ced12, dCed-12, dCed12, dced-12, dced12, elmo
Chromosome Location
Chr.2L: 12097838 - 12100839 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032409
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135704.3 NP_609548.1 1 - 2 225 84
GenBank mRNA AF417860.1 - 1 - 2 112 84
BT100121.1 - 1 - 2 221 84

View Full Product Details