Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RNaseZ
- Entrez Gene ID
-
36086
- Gene Name
- Ribonuclease Z
- Gene Aliases
-
CG3298, Dmel\CG3298, Dmel_CG3298, JHI-1, JhI-1, RNZ, dRNase Z, dRNaseZ, dtRNaseZ, jhI-1, tRNAse Z, tRNase Z, tRNase Z[L]
- Chromosome Location
-
Chr.2R: 10307030 - 10309794 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028426
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165763.3 |
NP_724916.1 |
1 - 2 |
1356 |
86 |
View Full Product Details