Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Spat
- Entrez Gene ID
-
31587
- Gene Name
- Serine pyruvate aminotransferase
- Gene Aliases
-
CG3926, Dm-Spat, Dmel\CG3926, Dmel_CG3926, dAGXT, spa, spat
- Chromosome Location
-
Chr.X: 6528948 - 6530726 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0014031
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078507.2 |
NP_511062.1 |
1 - 2 |
318 |
94 |
GenBank mRNA |
BT133279.1 |
- |
1 - 2 |
318 |
94 |
View Full Product Details