Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gip
Entrez Gene ID
31960
Gene Name
GIP-like
Gene Aliases
CG2227, Dmel\CG2227, Dmel_CG2227, GIP, T15, gene C
Chromosome Location
Chr.X: 10328877 - 10329872 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011770
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078551.4 NP_511106.1 1 - 1 871 57
GenBank mRNA AY071075.1 - 1 - 1 848 57
M18635.1 - - 1245 57

View Full Product Details