Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tim
Entrez Gene ID
33571
Gene Name
timeless
Gene Aliases
CG3234, Dmel\CG3234, Dmel_CG3234, Ritsu, TIM, Tim, Tim-1, dTIM, dTim, dtim, mel_tim, rit, s-tim, tim1
Chromosome Location
Chr.2L: 3493986 - 3508119 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014396
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014463.4 NP_001014463.2 9 - 10 2788 92
NM_001038783.3 NP_001033872.2 8 - 9 3215 92
NM_001144308.3 NP_001137780.2 9 - 10 2831 92
NM_001144309.4 NP_001137781.2 8 - 9 3270 92
NM_001201757.2 NP_001188686.2 8 - 9 2881 92
NM_001347756.1 NP_001334730.1 8 - 9 2884 92
NM_164540.4 NP_722912.3 8 - 9 3215 92
NM_164541.6 NP_722913.3 8 - 9 2888 92
NM_164542.5 NP_722914.3 8 - 9 2884 92

View Full Product Details