Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
msb1l
Entrez Gene ID
35254
Gene Name
CG10364-PA
Gene Aliases
CG10364, Dmel\CG10364, Dmel_CG10364, Msb1l, msh1l
Chromosome Location
Chr.2L: 19577701 - 19579131 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027949
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136147.4 NP_609991.1 1 - 2 272 96
GenBank mRNA AY051807.1 - 1 - 2 238 96

View Full Product Details