Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mtp
- Entrez Gene ID
-
35362
- Gene Name
- Microsomal triacylglycerol transfer protein
- Gene Aliases
-
2L3637, 2L4501, CG9342, CT3751, Dmel\CG9342, Dmel_CG9342, MTP, dMTP, mtp
- Chromosome Location
-
Chr.2L: 20919094 - 20922876 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0266369
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136231.4 |
NP_610075.2 |
1 - 2 |
194 |
61 |
View Full Product Details