Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
az2
Entrez Gene ID
35682
Gene Name
CG1605-PA
Gene Aliases
CG1605, Dmel\CG1605, Dmel_CG1605, zf43C
Chromosome Location
Chr.2R: 7510622 - 7512749 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025185
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136445.3 NP_610289.2 1 - 2 483 61
GenBank mRNA BT021301.1 - 1 - 2 458 61

View Full Product Details