Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lolal
Entrez Gene ID
44703
Gene Name
lola like
Gene Aliases
25/12, Ban, CG5738, Dmel\CG5738, Dmel_CG5738, EP0647, EP647, LOLAL, Lolal, anon-EST:fe3D3, anon-WO0172774.186, ban, bat, l(2)k02512, l(2)kO2512
Chromosome Location
Chr.2R: 18154095 - 18158192 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0022238
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169715.2 NP_001163186.1 2 - 3 577 68
NM_001259473.2 NP_001246402.1 3 - 4 475 68
NM_001259474.2 NP_001246403.1 3 - 4 509 68
NM_001299635.1 NP_001286564.1 3 - 4 462 68
NM_080039.4 NP_524778.1 3 - 4 440 68
NM_166270.2 NP_725756.1 3 - 4 583 68
NM_166271.3 NP_725757.1 3 - 4 536 68
NM_166272.3 NP_725758.1 3 - 4 500 68
GenBank mRNA AF308476.1 - 3 - 4 398 68
AY060364.1 - 3 - 4 396 68
BT099722.1 - 2 - 3 2159 68
KX531691.1 - 2 - 3 191 68

View Full Product Details