Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pepck1
- Entrez Gene ID
-
37131
- Gene Name
- Phosphoenolpyruvate carboxykinase 1
- Gene Aliases
-
143299_at, CG10924, CG17725, Dmel\CG17725, Dmel_CG17725, Dromel_CG17725_FBtr0086701_pepck_mORF, PEPCK, Pepck, dPEPCK, pepck, pepck1
- Chromosome Location
-
Chr.2R: 18536767 - 18539416 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003067
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079060.3 |
NP_523784.2 |
2 - 2 |
2384 |
120 |
GenBank mRNA |
BT003447.1 |
- |
2 - 2 |
2383 |
120 |
- Gene Symbol
- CG45087
- Entrez Gene ID
-
19834964
- Gene Name
- CG45087-PA
- Gene Aliases
-
Dmel\CG45087, Dmel_CG45087, Dromel_CG17725_FBtr0086701_pepck_uORF
- Chromosome Location
-
Chr.2R: 18536767 - 18539416 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0266488
View Full Product Details