Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr58c
- Entrez Gene ID
-
37501
- Gene Name
- Gustatory receptor 58c
- Gene Aliases
-
58A.3, 58A3, CG13491, Dmel\CG13491, Dmel_CG13491, GR58A.3, GR58c, Gr58A3
- Chromosome Location
-
Chr.2R: 21976662 - 21977957 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041237
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166489.2 |
NP_726133.2 |
1 - 2 |
1065 |
92 |
View Full Product Details