Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spel1
Entrez Gene ID
34842
Gene Name
spellchecker1
Gene Aliases
BG:DS01068.9, CG4215, Dmel\CG4215, Dmel_CG4215, Msh2, SPEL1, spel, spel-1
Chromosome Location
Chr.2L: 14362069 - 14366840 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015546
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014482.3 NP_001014482.2 5 - 6 2273 120
NM_001259102.2 NP_001246031.1 5 - 6 2273 120
NM_001299002.1 NP_001285931.1 5 - 6 2273 120
NM_078841.5 NP_523565.2 5 - 6 2273 120

View Full Product Details