Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vg
Entrez Gene ID
36421
Gene Name
vestigial
Gene Aliases
CG3830, Dmel\CG3830, Dmel_CG3830, VG, Vg, vg21
Chromosome Location
Chr.2R: 12884632 - 12899385 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003975
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078999.4 NP_523723.1 3 - 4 2037 103

View Full Product Details