Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spt
Entrez Gene ID
35889
Gene Name
Spitting Image
Gene Aliases
CG8247, Dmel\CG8247, Dmel_CG8247, SPT, spt
Chromosome Location
Chr.2R: 8920270 - 8922755 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033348
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136579.5 NP_610423.3 5 - 6 1727 95
GenBank mRNA AY071398.1 - 5 - 6 1723 95

View Full Product Details