Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hen1
- Entrez Gene ID
-
36301
- Gene Name
- Hen1 methyltransferase
- Gene Aliases
-
CG12367, DmHen1, Dmel\CG12367, Dmel_CG12367, FBgn 33686, PIMET, Pimet, Pimet/DmHen1, hen-1, hen1
- Chromosome Location
-
Chr.2R: 12145710 - 12152752 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033686
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136888.3 |
NP_610732.1 |
1 - 2 |
268 |
63 |
GenBank mRNA |
AY069325.1 |
- |
1 - 2 |
251 |
63 |
View Full Product Details