Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gr61a
Entrez Gene ID
38098
Gene Name
Gustatory receptor 61a
Gene Aliases
CG13888, Dmel61a, Dmel\CG13888, Dmel_CG13888, GR61a, Gr61D1
Chromosome Location
Chr.3L: 841451 - 843701 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035167
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274297.1 NP_001261226.1 4 - 5 970 100
NM_001299963.1 NP_001286892.1 4 - 5 966 100
NM_079153.4 NP_523877.1 4 - 5 970 100

View Full Product Details