Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ras64B
- Entrez Gene ID
-
38494
- Gene Name
- Ras oncogene at 64B
- Gene Aliases
-
BcDNA:RE36103, C-ras2, CG1167, D-ras-2, D-ras2, DRas2, Dm-ras-64B, Dmel\CG1167, Dmel_CG1167, Dmras64B, Dras2, Dras64B, RAS, RAS2, RAS64B, Ras, Ras2, c-ras2, ras, ras-2, ras2
- Chromosome Location
-
Chr.3L: 4139076 - 4141398 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003206
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079193.4 |
NP_523917.2 |
2 - 3 |
276 |
66 |
GenBank mRNA |
AY119135.1 |
- |
2 - 3 |
274 |
66 |
View Full Product Details