Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- bin
- Entrez Gene ID
-
38766
- Gene Name
- biniou
- Gene Aliases
-
BIN, Bin, CG18647, DmFoxF, Dmbin, Dmel\CG18647, Dmel_CG18647, FOXF, FoxF, XIV, binI, l(3)65CEe
- Chromosome Location
-
Chr.3L: 6990622 - 6994447 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0045759
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079226.3 |
NP_523950.2 |
1 - 2 |
1238 |
61 |
GenBank mRNA |
BT025075.1 |
- |
1 - 2 |
1218 |
61 |
View Full Product Details