Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptp69D
Entrez Gene ID
39443
Gene Name
Protein tyrosine phosphatase 69D
Gene Aliases
69D, CG10975, CT30751, DPTP, DPTP69D, Dmel\CG10975, Dmel_CG10975, Dptp69D, PTP69D, PTPase, Ptp, Ptp69d, R-PTP 69D, RPtp69D, dptp69D, dptp69d, ptp69D, ptp69d, rptp69d
Chromosome Location
Chr.3L: 12734945 - 12741568 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014007
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079324.3 NP_524048.2 6 - 7 3631 99
NM_176324.2 NP_788502.1 6 - 7 3628 99
GenBank mRNA BT001531.1 - 6 - 7 3624 99

View Full Product Details