Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bru3
Entrez Gene ID
39527
Gene Name
bruno 3
Gene Aliases
Bru-3, CG10046, CG12478, CG34243, CG43744, Dm Bru-3, Dmel\CG43744, Dmel_CG43744, bru-3, bruno-3, cg10046, cg12478, l(3)05871
Chromosome Location
Chr.3L: 13521907 - 13803400 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264001
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202161.2 NP_001189090.1 8 - 9 1542 67
NM_001202162.2 NP_001189091.1 7 - 8 1500 67
NM_001202163.2 NP_001189092.1 7 - 8 1500 67
NM_001202164.2 NP_001189093.1 6 - 7 1365 67
NM_001202165.2 NP_001189094.1 7 - 8 1524 67
NM_001202166.4 NP_001189095.1 7 - 8 1524 67
NM_001202167.2 NP_001189096.1 7 - 8 1506 67
NM_001202168.2 NP_001189097.1 7 - 8 1819 67
NM_001202169.2 NP_001189098.1 6 - 7 1365 67
NM_001202170.2 NP_001189099.1 6 - 7 1389 67
NM_001274878.1 NP_001261807.1 9 - 10 1884 67
NM_001274879.1 NP_001261808.1 9 - 10 1620 67
NM_001382115.1 NP_001369049.1 7 - 8 1500 67
NM_001382116.1 NP_001369050.1 7 - 8 1500 67
NM_001382117.1 NP_001369051.1 8 - 9 1485 67
NM_079335.6 NP_524059.4 6 - 7 1365 67
NM_168557.3 NP_729923.1 8 - 9 1542 67
GenBank mRNA AY060398.1 - 8 - 9 1542 67

View Full Product Details