The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202163.2 | NP_001189092.1 | 8 - 9 | 1601 | 70 |
NM_001202166.4 | NP_001189095.1 | 8 - 9 | 1625 | 70 | |
NM_001202171.2 | NP_001189100.1 | 7 - 8 | 1487 | 70 | |
NM_001274878.1 | NP_001261807.1 | 10 - 11 | 1985 | 70 | |
NM_001274879.1 | NP_001261808.1 | 10 - 11 | 1721 | 70 | |
NM_001382115.1 | NP_001369049.1 | 8 - 9 | 1601 | 70 | |
NM_001382116.1 | NP_001369050.1 | 8 - 9 | 1601 | 70 | |
NM_001382117.1 | NP_001369051.1 | 9 - 10 | 1586 | 70 | |
NM_079335.6 | NP_524059.4 | 7 - 8 | 1466 | 70 | |
NM_168557.3 | NP_729923.1 | 9 - 10 | 1643 | 70 | |
GenBank mRNA | AY060398.1 | - | 9 - 10 | 1643 | 70 |