Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ftz-f1
- Entrez Gene ID
-
40045
- Gene Name
- ftz transcription factor 1
- Gene Aliases
-
BFTZ-F1, CG13378, CG4059, DEP1, DmFTZ-F1, Dmel\CG4059, Dmel_CG4059, FIP1, FTZ-F1, FTZ-F1A, FTZ-F1alpha, FTZ-Fl, FTZF1, FTZf1alpha, Ftz-F1, Ftz-f1, Ftzf1, NR5A3, alphaFTZ-F1, alphaFTZF1, alphaftz-f1, bFTZ-F1, beta FTZ-F1, beta Ftz-f1, beta-FTZ-F1, beta-Ftz-F1, betaFTZ-F1, betaFTZF1, betaFtz-F1, betaFtz-f1, betaftz-F1, betaftz-f1, betaftzf1, dFTZ-F1alpha, dFtz-F1alpha, dmFTZ-f1, ftz-F1, ftz-f1alpha, ftzf1, l(3)03649
- Chromosome Location
-
Chr.3L: 18750607 - 18801309 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001078
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079419.3 |
NP_524143.2 |
2 - 3 |
2522 |
62 |
GenBank mRNA |
BT058016.1 |
- |
2 - 3 |
2108 |
62 |
View Full Product Details