Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RpS23
Entrez Gene ID
36576
Gene Name
Ribosomal protein S23
Gene Aliases
CG8415, CT24703, Dmel\CG8415, Dmel_CG8415, RPS23, Rp S23, anon-EST:Posey158, anon-EST:Posey63
Chromosome Location
Chr.2R: 14215096 - 14216627 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033912
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299481.1 NP_001286410.1 2 - 3 212 69
NM_137095.3 NP_610939.2 2 - 3 261 69
GenBank mRNA AY089523.1 - 2 - 3 238 69
KX532076.1 - 1 - 2 167 69

View Full Product Details