Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tomosyn
Entrez Gene ID
32217
Gene Name
CG17762-PC
Gene Aliases
BcDNA:RH19475, CG11056, CG17762, CG2205, Dmel\CG17762, Dmel_CG17762, tomosyn
Chromosome Location
Chr.X: 12631605 - 12653442 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030412
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169264.2 NP_001162735.1 3 - 4 862 62
NM_001169265.2 NP_001162736.2 3 - 4 862 62
NM_001258715.2 NP_001245644.1 3 - 4 862 62
NM_132591.5 NP_572819.4 4 - 5 907 62
NM_167328.4 NP_727628.3 4 - 5 902 62
NM_167329.3 NP_727629.2 3 - 4 1062 62
NM_167330.3 NP_727630.3 3 - 4 984 62

View Full Product Details