Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lip2
Entrez Gene ID
43980
Gene Name
Lipase 2
Gene Aliases
CG17116, DILP2, Dmel\CG17116, Dmel_CG17116, Dmlip2, dilp2, lip2
Chromosome Location
Chr.2L: 10701940 - 10709453 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024740
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298883.1 NP_001285812.1 2 - 3 771 77
NM_079928.2 NP_524667.1 2 - 3 284 77

View Full Product Details