Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trbl
Entrez Gene ID
43999
Gene Name
tribbles
Gene Aliases
CG5408, Dmel\CG5408, Dmel_CG5408, Trbl, trb
Chromosome Location
Chr.3L: 20396288 - 20401609 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028978
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079933.4 NP_524672.1 1 - 2 1183 75
GenBank mRNA AF204688.1 - 1 - 2 1161 75
BT004834.1 - 1 - 2 1184 75

View Full Product Details