Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ste24b
- Entrez Gene ID
-
36907
- Gene Name
- ste24b prenyl protease type I
- Gene Aliases
-
CG9001, Dmel\CG9001, Dmel\Ste24, Dmel\Ste24b, Dmel_CG9001
- Chromosome Location
-
Chr.2R: 16998610 - 17000126 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034175
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137330.1 |
NP_611174.1 |
1 - 2 |
129 |
123 |
View Full Product Details