Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp12b2
- Entrez Gene ID
-
37163
- Gene Name
- CG15077-PA
- Gene Aliases
-
12b2, CG15077, Cyp12B2, Dmel\CG15077, Dmel_CG15077
- Chromosome Location
-
Chr.2R: 18754995 - 18757083 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034387
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137526.3 |
NP_611370.2 |
3 - 4 |
588 |
87 |
View Full Product Details