Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
miple2
Entrez Gene ID
44787
Gene Name
midkine and pleiotrophin 2
Gene Aliases
CG18321, Dmel\CG18321, Dmel_CG18321, Miple2
Chromosome Location
Chr.3L: 271200 - 275018 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029002
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274270.1 NP_001261199.1 3 - 4 777 65
NM_001274271.1 NP_001261200.1 3 - 4 670 65
NM_001274272.1 NP_001261201.1 3 - 4 931 65
NM_143739.3 NP_651996.2 3 - 4 777 65
GenBank mRNA BT044311.1 - 3 - 4 775 65

View Full Product Details