Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
JYalpha
Entrez Gene ID
5740846
Gene Name
CG45760-PB
Gene Aliases
CG17923, CG40625, CG45760, Dmel\CG45760, Dmel_CG45760, JY-alpha, JYAlpha, Jyalpha
Chromosome Location
Chr.4: 879 - 5039 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267363
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110622.3 NP_001104092.2 4 - 5 2728 123

View Full Product Details