Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCB7
Entrez Gene ID
38193
Gene Name
ATP binding cassette subfamily B member 7
Gene Aliases
CG7955, Dmel\CG7955, Dmel_CG7955, dABCB7
Chromosome Location
Chr.3L: 1618414 - 1623413 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035244
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_167902.2 NP_728642.2 1 - 2 198 83
NM_167903.2 NP_728643.2 1 - 2 198 83

View Full Product Details