Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4s3
Entrez Gene ID
32444
Gene Name
CG9081-PA
Gene Aliases
4s3, CG9081, Dmel\CG9081, Dmel_CG9081, RE63964
Chromosome Location
Chr.X: 15113302 - 15116542 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030615
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_132775.3 NP_573003.2 2 - 3 780 70
GenBank mRNA BT133503.1 - 2 - 3 777 70

View Full Product Details