Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mef2
Entrez Gene ID
36032
Gene Name
Myocyte enhancer factor 2
Gene Aliases
22.21, BEST:SD04091, C, CG1429, D-MEF2, D-Mef2, D-mef2, DMEF-2, DMEF2, DMedf2, DMef-2, DMef2, Dmef, Dmef-2, Dmef2, Dmel\CG1429, Dmel_CG1429, MEF-2, MEF2, Mef, Mef-2, SD04091, dMEF2, dMEf2, dMef2, dmef2, l(2)46CFr, mb247, mef, mef-2, mef2, mef2c
Chromosome Location
Chr.2R: 9913496 - 9958858 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011656
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382029.1 NP_001369071.1 8 - 9 2457 55
NM_057673.5 NP_477021.1 8 - 9 2439 55
GenBank mRNA X83527.1 - - 2421 55

View Full Product Details