Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cht4
- Entrez Gene ID
-
49815
- Gene Name
- Chitinase 4
- Gene Aliases
-
BcDNA:RE62779, CG3986, CHT4, DmCHT4, DmCht4, Dmel\CG3986, Dmel_CG3986
- Chromosome Location
-
Chr.2R: 21065379 - 21067087 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0022700
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080223.3 |
NP_524962.2 |
2 - 3 |
270 |
73 |
GenBank mRNA |
AY071559.1 |
- |
2 - 3 |
271 |
73 |
View Full Product Details