Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rbf
- Entrez Gene ID
-
31027
- Gene Name
- Retinoblastoma-family protein
- Gene Aliases
-
CG7413, Dmel\CG7413, Dmel_CG7413, EG:34F3.3, FBF, RB, RBF, RBF1, RBf1, Rb, Rb1, RbF, Rbf1, dRBF, pRb, rb, rbf, rbf1
- Chromosome Location
-
Chr.X: 797090 - 801479 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015799
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080297.3 |
NP_525036.2 |
5 - 6 |
1898 |
70 |
GenBank mRNA |
AY089532.1 |
- |
5 - 6 |
1876 |
70 |
View Full Product Details