Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SK
Entrez Gene ID
31456
Gene Name
small conductance calcium-activated potassium channel
Gene Aliases
BcDNA:GM01744, CG10706, CG12733, CG4179, Dmel\CG10706, Dmel_CG10706, Sk, dSK
Chromosome Location
Chr.X: 5340236 - 5405138 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029761
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297959.1 NP_001284888.1 12 - 13 2294 61
NM_080339.3 NP_525078.3 12 - 13 2294 61

View Full Product Details