Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- aft
- Entrez Gene ID
-
37034
- Gene Name
- adrift
- Gene Aliases
-
CG5032, Dmel\CG5032, Dmel_CG5032, EG:52C10.7
- Chromosome Location
-
Chr.2R: 17786135 - 17788556 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026309
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058065.3 |
NP_477413.1 |
2 - 3 |
553 |
110 |
View Full Product Details