Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Taz
- Entrez Gene ID
-
36405
- Gene Name
- Tafazzin
- Gene Aliases
-
CG8766, Dmel\CG8766, Dmel_CG8766, TAZ, dTAZ, tafazzin, taz
- Chromosome Location
-
Chr.2R: 12753026 - 12756414 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026619
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058084.5 |
NP_477432.3 |
1 - 2 |
479 |
69 |
GenBank mRNA |
AY071059.1 |
- |
1 - 2 |
476 |
69 |
View Full Product Details