Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mbc
Entrez Gene ID
42817
Gene Name
myoblast city
Gene Aliases
CG10379, DOCK180, Dm MBC, Dmel\CG10379, Dmel_CG10379, Dock180, MBC, MBC/DOCK180, Mbc, Su(rac)1, dock180
Chromosome Location
Chr.3R: 23781765 - 23801634 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015513
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260338.2 NP_001247267.1 5 - 6 2470 93
NM_057796.4 NP_477144.1 5 - 6 2470 93
GenBank mRNA AF007805.1 - 5 - 6 2455 93

View Full Product Details