Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mfas
Entrez Gene ID
41455
Gene Name
midline fasciclin
Gene Aliases
87A7-9/7, BEST:GH06752, CG3359, Dmel\CG3359, Dmel_CG3359, Mfas, anon-87Af, anon-EST:Liang-97, anon-WO0140519.174, clone 97, gh06752
Chromosome Location
Chr.3R: 11984862 - 11993315 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260745
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260137.1 NP_001247066.1 2 - 3 277 99
NM_079600.3 NP_524324.2 3 - 4 607 99
NM_169444.1 NP_731661.1 3 - 4 295 99
NM_176466.1 NP_788643.1 3 - 4 314 99
NM_176467.1 NP_788644.1 3 - 4 292 99
NM_176468.1 NP_788645.1 2 - 3 691 99
NM_176469.2 NP_788646.1 3 - 4 412 99
NM_176470.1 NP_788647.1 3 - 4 317 99
NM_176471.1 NP_788648.1 3 - 4 490 99
NM_176472.1 NP_788649.1 3 - 4 512 99
NM_176473.1 NP_788650.1 4 - 5 386 99
NM_176474.1 NP_788651.1 4 - 5 364 99
NM_176475.1 NP_788652.1 3 - 4 763 99
NM_176476.1 NP_788653.1 4 - 5 367 99
NM_176477.1 NP_788654.1 4 - 5 389 99
NM_176478.1 NP_788655.1 4 - 5 562 99
GenBank mRNA AY069094.1 - 3 - 4 277 99
BT120190.1 - 2 - 3 277 99
BT125878.1 - 4 - 5 363 99
BT133296.1 - 4 - 5 541 99

View Full Product Details