Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rtet
- Entrez Gene ID
-
42500
- Gene Name
- tetracycline resistance
- Gene Aliases
-
BcDNA:LD28419, CG5760, Dmel\CG5760, Dmel_CG5760, Drtet, anon-WO0119857.1, l(3)07086
- Chromosome Location
-
Chr.3R: 21108446 - 21110834 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028468
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079705.3 |
NP_524429.1 |
2 - 3 |
944 |
69 |
GenBank mRNA |
AF181656.1 |
- |
2 - 3 |
896 |
69 |
View Full Product Details