Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hph
Entrez Gene ID
40633
Gene Name
HIF prolyl hydroxylase
Gene Aliases
0303/04, 31543, CG1114, CG14665, CG2676, CG31543, CG44015, CR31541, Dm.egl-9, Dmel\CG44015, Dmel_CG44015, Fga, FgaB, PHD, anon-EST:Posey69, dHPH, dPHD, dPHD2, dmHPH, fatiga, fga, hph, l(3)02255, l(3)82Fe, l(3)S030304
Chromosome Location
Chr.3R: 5257037 - 5269507 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264785
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300242.1 NP_001287171.1 4 - 5 1591 70
NM_141268.3 NP_649525.1 4 - 5 1205 70
NM_169036.2 NP_730906.1 4 - 5 1591 70
NM_169037.3 NP_730908.1 4 - 5 770 70
GenBank mRNA AY051594.1 - 4 - 5 836 70
AY094896.1 - 4 - 5 1587 70
BT001659.1 - 4 - 5 765 70

View Full Product Details