Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osi11
Entrez Gene ID
40765
Gene Name
Osiris 11
Gene Aliases
CG15596, Dmel\CG15596, Dmel_CG15596, osi11
Chromosome Location
Chr.3R: 6267591 - 6268757 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037418
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141373.2 NP_649630.1 1 - 1 919 60
GenBank mRNA BT100070.2 - 1 - 1 850 60

View Full Product Details