Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WASp
Entrez Gene ID
43402
Gene Name
CG1520-PA
Gene Aliases
CG1520, D-WASP, Dm WASP, DmWASP, Dmel\CG1520, Dmel_CG1520, WASP, WSP, Wasp, Wsp, dWASP, wasp, wsp
Chromosome Location
Chr.3R: 28824506 - 28832024 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024273
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276116.1 NP_001263045.1 4 - 5 1185 78
NM_143380.4 NP_651637.1 4 - 5 1245 78
NM_170378.2 NP_733257.1 4 - 5 1133 78
NM_176578.2 NP_788755.1 3 - 4 951 78
GenBank mRNA AY070576.1 - 4 - 5 1256 78

View Full Product Details