Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UbcE2H
Entrez Gene ID
31728
Gene Name
Ubiquitin conjugating enzyme E2H
Gene Aliases
CG2257, Dmel\CG2257, Dmel_CG2257, Ubc-2EH, Ubc-E2H, Ubc-E2Hs, UbcDE2H, dUbc-E2H, ubcE2h
Chromosome Location
Chr.X: 8087544 - 8094064 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029996
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_132210.3 NP_572438.1 2 - 2 2185 147
NM_167145.2 NP_727233.1 2 - 2 2322 147
NM_167146.2 NP_727234.1 2 - 2 2221 147
GenBank mRNA AY061237.1 - 2 - 2 2164 147

View Full Product Details