Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ccp84Ab
- Entrez Gene ID
-
40824
- Gene Name
- CG1252-PA
- Gene Aliases
-
BG:DS00004.2, CG1252, Ccp, DmelCcp84Ab, Dmel\CG1252, Dmel_CG1252, cc7
- Chromosome Location
-
Chr.3R: 6701968 - 6702877 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004782
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141425.2 |
NP_649682.1 |
2 - 2 |
740 |
113 |
View Full Product Details