Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Set1
Entrez Gene ID
3354971
Gene Name
SET domain containing 1
Gene Aliases
CG17395, CG17396, CG40351, Dmel\CG40351, Dmel_CG40351, SET1, SETD1, aaf45425 Dm, dSET1, dSet1, dset1, l(3)h5, l5, set1
Chromosome Location
Chr.3L: 25107594 - 25115180 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040022
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015221.3 NP_001015221.1 5 - 6 4845 133
NM_001015222.3 NP_001015222.1 5 - 6 5028 133
NM_001110936.3 NP_001104406.1 5 - 6 5037 133
NM_001170375.2 NP_001163846.1 6 - 7 4853 133
NM_001170376.2 NP_001163847.1 5 - 6 4868 133
NM_001170377.1 NP_001163848.1 5 - 6 4832 133
NM_001170378.1 NP_001163849.1 5 - 6 4821 133
NM_001170379.1 NP_001163850.1 4 - 5 4818 133
NM_001170380.2 NP_001163851.1 5 - 6 4859 133

View Full Product Details