Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prdm8
Entrez Gene ID
406719
Gene Name
PR domain containing 8
Gene Aliases
prdm8a, wu:fd12b11, zgc:63680
Chromosome Location
Chr.13: 31292699 - 31296494 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213410.1 NP_998575.1 3 - 4 608 81
XM_005156810.2 XP_005156867.1 3 - 4 748 81
GenBank mRNA BC055201.1 - 3 - 4 608 81

View Full Product Details