Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dipk2aa
Entrez Gene ID
492795
Gene Name
divergent protein kinase domain 2Aa
Gene Aliases
c3orf58a, dia1a, zgc:101666
Chromosome Location
Chr.2: 8261241 - 8312199 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007436.1 NP_001007437.1 3 - 4 1332 64
GenBank mRNA BC085407.1 - 3 - 4 1332 64

View Full Product Details