Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eomesb
Entrez Gene ID
799447
Gene Name
eomesodermin homolog b
Gene Aliases
Eomes2
Chromosome Location
Chr.16: 55168245 - 55182394 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083575.2 NP_001077044.1 6 - 7 1255 118
GenBank mRNA AB269787.1 - 6 - 7 1256 118

View Full Product Details